Learn More
Mopec Stationery Book Stand
SDP
Catalog No. 23900717 Shop All Mopec Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
23-900-717 Stainless Steel
1 options
Catalog No. 23-900-717 Supplier Mopec Supplier No. BN200
Add to Cart
Add to Cart
Built for laboratories, pathology suites, and educational environments, offers a versatile reference podium that feature a unique dual-sided book rest.
  • Heliarc-welded seams and joints, ground and polished to match adjoining surfaces
  • 304 stainless steel construction with a polished No. 4 finish
  • (2) threaded knobs for adjusting height
  • (2) 3.5 in. swivel front casters and (2) 3.5 in. locking swivel rear casters
  • BN100: Mobile
  • BN200: Stationary
Specifications
Product Type Book Truck
Material Stainless Steel
No. of Shelves 2