Learn More
Electron Microscopy Sciences StatMark Pen
SDP
Catalog No. 50365873
Encompass_Preferred
Permanent ink in an ultrafine tip is ideal for labeling microscope slides and cassettes. StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.
Catalog No. 50-365-873 Supplier Electron Microscopy Sciences Supplier No. 7210912
Permanent ink in an ultrafine tip is ideal for labeling microscope slides and cassettes. StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.