Learn More
Sigma Aldrich Fine Chemicals Biosciences STATMARK permanent marker
Catalog No. 502901650
Encompass
The STATMARK Permanent ink pen in an ultra fine tip is ideal for labeling microscope slides and cassettes. Made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
Catalog No. 50-290-1650 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z64819112EA
The STATMARK Permanent ink pen in an ultra fine tip is ideal for labeling microscope slides and cassettes. Made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.