Learn More
Tri Continent Scientific StatMatic Ii, 8-Channel Microplate Washer And Bottle Top Dispenser
Catalog No. NC0610470
Encompass_Preferred
Statmatic Ii - 8-Channel Microplate Washer And Bottle Top Dispenser. Ideal For Hand-Held Applications. The Statmatic Ii Microplate Washer And Bottle Top Dispenser With A Hand-Held Eight-Port Disposable Manifold Provides Greater Ease, Speed, Convenience, And Affordability Than A Multi-Channel Pipette. The Statmatic Ii Microplate Washer And Bottle Top Dispenser Can Be Connected To A Range Of Bottle Sizes And Can Completely Fill A Microplate With One-Handed Operation -In A Matter Of Seconds. Eight-Port Manifold, Designed For 96-Well Microplates, Delivers Fluids In Volumes Of 100, 200, 250, Or 300 l With Well-To-Well Accuracy Of 5% Or Better. Constructed Of Polypropylene, Teflon, Glass, And Ceramics, There Is No Exposed Glass That Can Break And Requires Minimal Maintenance. Compatible With Most Reagents And Chemicals And Designed So That Even Protein-Based Reagents Won'T Cause Sticking. Filtered Air Vent Keeps Liquids Free Of Contaminants.
Catalog No. NC0610470 Supplier Tri Continent Scientific Supplier No. 0980
Statmatic Ii - 8-Channel Microplate Washer And Bottle Top Dispenser. Ideal For Hand-Held Applications. The Statmatic Ii Microplate Washer And Bottle Top Dispenser With A Hand-Held Eight-Port Disposable Manifold Provides Greater Ease, Speed, Convenience, And Affordability Than A Multi-Channel Pipette. The Statmatic Ii Microplate Washer And Bottle Top Dispenser Can Be Connected To A Range Of Bottle Sizes And Can Completely Fill A Microplate With One-Handed Operation -In A Matter Of Seconds. Eight-Port Manifold, Designed For 96-Well Microplates, Delivers Fluids In Volumes Of 100, 200, 250, Or 300 l With Well-To-Well Accuracy Of 5% Or Better. Constructed Of Polypropylene, Teflon, Glass, And Ceramics, There Is No Exposed Glass That Can Break And Requires Minimal Maintenance. Compatible With Most Reagents And Chemicals And Designed So That Even Protein-Based Reagents Won'T Cause Sticking. Filtered Air Vent Keeps Liquids Free Of Contaminants.