Learn More
Waters Corp Waters Stator Face Seal Assembly, 7725i
Catalog No. 50793663 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Stator Face Seal Assembly, 7725i, creates a high-pressure seal between the stator and the alumina stator face. This assembly includes a stator seal disc that connects the six stator ports to the six corresponding holes in the stator face.
  • Includes a stator seal disc
  • Creates a high-pressure seal between the stator and alumina stator face
  • Connects six stator ports to six matching holes in the stator face
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Replacement schedule: As required
Catalog No. 50-793-663 Supplier Waters Corp Supplier No. WAT055947
The Stator Face Seal Assembly, 7725i, creates a high-pressure seal between the stator and the alumina stator face. This assembly includes a stator seal disc that connects the six stator ports to the six corresponding holes in the stator face.
  • Includes a stator seal disc
  • Creates a high-pressure seal between the stator and alumina stator face
  • Connects six stator ports to six matching holes in the stator face
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Replacement schedule: As required