Learn More
Trajan Scientific VICI Valco Stator, 6 Port, 15K-20K PSI, 0.15mm Bore, For C72VU-6676D-CTC
Catalog No. NC1264325
Encompass
VICI Valco Stator, 6 Port, 15K-20K PSI, 0.15mm Bore, Cheminert, vertical Injection For Valves:VAL.C72VX-6676D-CTCVAL.C72VU-6676D-CTC Thermo Part Number: 00109-99-00009. Made in the USA.
Catalog No. NC1264325 Supplier Trajan Scientific Supplier No. VAL.C72V6C76
VICI Valco Stator, 6 Port, 15K-20K PSI, 0.15mm Bore, Cheminert, vertical Injection For Valves:VAL.C72VX-6676D-CTCVAL.C72VU-6676D-CTC Thermo Part Number: 00109-99-00009. Made in the USA.