Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desco™ Statshield™ Metal-In Zip Bag

Click to view available options
Dimensions (L x W):
5 x 3 in.
6 x 4 in.
8 x 6 in.
10 x 8 in.
12 x 10 in.
Description
- Most commonly used shielding bag meets limits of ANSI/ESD S541 per ANSI/ESD S20.20
- High volume manufacturing provides value for the customer
- Integral antistatic, low tribocharging properties
- Bag contents will not electrostatically charge during movement
- Integral static dissipative properties
- Not topically treated, not humidity dependant, won't wear off
- Manufactured on specially designed soft fold converting machines, which prevents scratches and creases maintaining the integrity of faraday cage shielding protection
- Greater sealing surface provides greater seal strength compared to comparable products with 0.125 in. wide seals
- Identified with ESD Protective Symbol as required by ANSI/ESD S541 with lot code number
- Improved quality control tracking and identified as ESD packaging
- Polycarbonate compatible containing no amines or N-octanic acid
- Non-corrosive, will not contaminate parts
- Testable to industry standards
- Meets or exceeds one or more of the following standards: electrical and physical requirements of ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, EIA 625, MIL-STD-1686, and EIA 583
- Low charging antistatic high visibility yellow reclosable zipper
Specifications
Specifications
| Color | Silver |
| For Use With (Application) | ESD sensitive devices |
| Material | Aluminized Polyester |
| Quantity | 100/Pk. |
| Dimensions (L x W) | 8 x 6 in. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction