Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Statshield™ Metal-Out Bag
SDP

Catalog No. 18688051
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-688-051 14 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-051 Supplier Desco™ Supplier No. 13275
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aluminum metal outer layer of laminated film features 40% light transmission, which allows for identification of bag contents.

  • Integral antistatic, low tribocharging properties
  • Bag contents will not electrostatically charge during movement
  • Integral static dissipative properties
  • Not topically treated, not humidity dependant, won't wear off
  • Manufactured on specially designed soft fold converting machines
  • Large wide seals provide high performance interior sealing layer
  • Identified with ESD Protective Symbol as required by ANSI/ESD S541
  • Improved quality control tracking and identified as ESD packaging
  • Lot coded
  • Quality control traceability
  • Polycarbonate compatible containing no amines or N-octanic acid
  • Non-corrosive, will not contaminate parts
  • Testable to industry standards
  • Antistatic, low charging, reclosable zipper
  • Lead-free RoHS compliant

Specifications

Autoclavable Not Autoclavable
Closure Type Zip Closure
Color Silver
For Use With (Application) ESD sensitive devices
Material Aluminized Polyester
Quantity 100/Pk.
Dimensions (L x W) 14 x 10 in.
Thickness (Metric) 0.070 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.