This Statshield Smock/Jacket with Knitted Cuffs offers robust ESD protection. It features an antistatic, low tribocharging design, creating a Faraday Cage effect to protect ESD-susceptible items. Panel-to-panel conductivity prevents isolated charged conductors. Meeting ANSI/ESD S20.20-2014 (< 3.5 X 107 ohms) as a Groundable Static Control System, it allows grounding of operators. It incorporates ≥9% carbon conductive nylon fibers for continuous charge dissipation, plus Hip-To-Cuff Grounding and two sleeve pen pockets.
- Antistatic, low tribocharging design
- Faraday cage effect for ESD-sensitive items
- Panel-to-panel conductivity
- Groundable static control garment (ANSI/ESD S20.20-2014 compliant)
- Carbon conductive nylon (≥9%) for consistent dissipation
- Hip-to-cuff grounding (no wrist cord needed)
- Two sleeve pen pockets