Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Statshield™ Smock Jacket with Knitted Cuffs
SDP

Catalog No. 18688178
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Color:
Black
Blue
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
18-688-178 Large Black
18-688-171 Small Blue
18-688-172 Medium Blue
18-688-173 Large Blue
18-688-174 X-Large Blue
18-688-175 2X-Large Blue
18-688-179 X-Large Black
18-688-180 2X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-688-178 Supplier Desco™ Supplier No. 73863
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Smock with groundable static control garment system that allows operators and smock to be bonded or electrically connected to ground to prevent ESD events in ESD protected areas.

  • Groundable static control garment system tested Per ANSI/ESD S20.20-2014 (Rtg < 3.5 X 107 ohms) Requirements, per ANSI/ESD STM2.1
  • Minimum of 9% carbon conductive nylon fibers woven in chain-link design provides continuous and consistent charge dissipation throughout lifetime of smock
  • Creates faraday cage effect around torso and arms of operator
  • Shields ESD charges from operators clothing from damaging ESD sensitive devices
  • Panel to panel conductivity
  • Prevents isolated charged conductor in ESD protected area
  • Hip-to-cuff grounding
  • Ground operator with no need for a cord to be attached to the operator's wrist
  • Two sleeve pen pockets
  • Holds pens or small tools in a safe and convenient location

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Material Polyester
Color Black
Closure Type Snaps
Cuff Style Knit
No. of Pockets 5
For Use With (Application) Prevent ESD Events in ESD Protected Areas
Length (Metric) 78.74 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.