Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America StatSpin™ CytoFuge™ 2 Personal Cytocentrifuge System

Catalog No. 220011
Change view
Click to view available options
Max. RCF:
1,060 x g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. RCF
22-001-1 1,060 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-001-1 Supplier HemoCue America Supplier No. CF02

missing translation for 'validMainframeMsg'

Includes

Centrifuge, 100/240V power supply, four-place rotor with cover, four filter concentrators with clips, four 1-well cell concentrators with gaskets, four 3-well cell concentrators with gaskets, 10 cell concentrator clips, four cell concentrator backing plates

Concentrates cells from body fluids onto microscope slides with a level of cell recovery, clarity and detail that cannot be achieved with direct smear techniques

Consistent samples are processed quickly and quietly; monolayer cell presentations preserve excellent morphological detail. Offers a choice of filter- or gasket-seal concentrators to handle all specimens from routine body fluid preparations to multiple cell concentrations or controls on the same slide. The broad range of selectable speeds and run times allow precise reproduction of most procedures.

Operating Features

  • Maximum speed/RCF: 4000rpm/1060×G
  • Intuitive speed and cycle time controls
  • Rotor included
  • Cover interlock prevents operation if cover is not completely closed; automatically releases upon cycle completion
  • Automatic acceleration and braking rates assure rapid, gentle starts and stops
  • Quiet-running and compact to fit any hood or benchtop space
  • Unit and rotor can be easily cleaned with bleach

Rotor Features
  • Requires no trunnion carriers or clamps
  • High-strength chassis and unbreakable see-through lid ensure mechanical containment
  • Leak-resistant and gasketed for improved biohazard containment

Order Info

Replacement clips, bags, filter cards, gaskets and backing plates all available separately.

TRUSTED_SUSTAINABILITY

Specifications

Max. RCF 1,060 x g
Max. Speed 4,000 rpm
Refrigerated No
Profile (Acceleration/Braking) Automatic acceleration and braking rates
Noise Level Quiet
Electrical Requirements 110/240 V 50/60 Hz
Height (English) Exterior 4.8 in.
Height (Metric) Exterior 12 cm
Length (English) Exterior 8.6 in.
Length (Metric) Exterior 19.3 cm
Width (English) Exterior 6.8 in.
Width (Metric) Exterior 16.3 cm
Dimensions (HWD) 4.8 x 6.8 x 8.6 in. (12 x 16.3 x 19.3 cm)
Run Time 2, 4, 6, 8, 10, 30 min. operating times
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.