Learn More
HemoCue America Slide Holders for StatSpin™ DiffSpin™ 2 Slide Spinner
Catalog No. 22314143
Change view
Click to view available options
Quantity:
100/Pk.
1000/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-314143 100/Pk.
22-020299 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-314143 Supplier HemoCue America Supplier No. SC01
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Slide holders for use with the DiffSpin™ 2 Slide Spinner. Easy to use slide holders help reduce biohazard risk.
DiffSpin disposable microscope slide holders
TRUSTED_SUSTAINABILITY
Specifications
Includes Microscope slides, slide holders and 50μL drop size transfer pipettes
For Use With (Equipment) DiffSpin 2 Slide Spinner (DS02)
For Use With (Application) For technique-independent blood films
Quantity 100/Pk.
Product Type Disposable Slide Holders