Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America StatSpin™ Express 2 Centrifuge

Catalog No. 22315792
Change view
Click to view available options
Max. Speed:
8,500 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. Speed
22-315-792 8,500 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-315-792 Supplier HemoCue America Supplier No. SSX2

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Includes

Unit includes four-place rotor, universal power supply, removable chamber liner, four 5mL inserts and North American power cord.

Ideal for rapid PPP for coagulation tests, sample prep for stat chemistry, cardiac profiles for the ER, sharp separations for send-out, and anywhere rapid serum or plasma is needed

  • Produces complete blood separations in 3 min. or less
  • Quiet and compact
  • Simple operation: Just load samples, select desired program and press START
  • Preprogrammed cycles eliminate confusion about speed and run time selection
  • Microprocessor control prevents overshoot and maintains speed and time to within 5%
  • “Incomplete Separation” alert ensures consistent results
  • Cover interlock system, chamber gasket and mechanical containment provide operator safety
  • Surfaces are 10% bleach-compatible
  • Polypropylene rotor makes cleaning easy

TRUSTED_SUSTAINABILITY

Specifications

Max. RCF 4,440 x g
Max. Speed 8,500 rpm
Refrigerated No
Noise Level Quiet
Profile (Acceleration/Braking) 20 sec. to 95% of speed / <30 sec.
Program Storage Preprogrammed
Rotor Included No
Run Time 30 to 180 sec.
Dimensions (HWD) 6 x 6.6 x 8.6 in. (15.2 x 16.8 x 21.8 cm)
Length (English) Exterior 8.6 in.
Length (Metric) Exterior 21.8 cm
Width (English) Exterior 6.6 in.
Width (Metric) Exterior 16.8 cm
Height (English) Exterior 6 in.
Height (Metric) Exterior 15.2 cm
Net Weight 5 lb. (2.3 kg)
Electrical Requirements 100/240 V 50/60 Hz
Amperage 1.7 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.