Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America StatSpin™ Microhematocrit Rotor

Catalog No. 22272546
Change view
Click to view available options
Capacity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-272546 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-272546 Supplier HemoCue America Supplier No. RH12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Twelve-place microhematocrit rotor with transparent lid

  • Accepts up to 12 StatSpin glass or plastic capillary tubes (40mm)
  • Includes carbon-fiber retaining ring for durability and easy-to-use transparent screw-on lid

TRUSTED_SUSTAINABILITY

Specifications

Type Microhematocrit Rotor
Capacity 5 mL
For Use With (Equipment) StatSpin MP or CritSpin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.