Learn More
HemoCue America StatSpin™ VET Express Centrifuge Adapters
Catalog No. 22269325
Change view
Click to view available options
For Use With (Equipment):
2mL BD Microtainer Tubes; Express 2 and 3 Centrifuge
3mL Tubes (10.25 x 64mm); Express 2 and 3 Centrifuge
5mL Tubes; Express 2 Centrifuge
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-269-325 2mL BD Microtainer Tubes; Express 2 and 3 Centrifuge
22-269283 5mL Tubes; Express 2 Centrifuge
22-269284 3mL Tubes (10.25 x 64mm); Express 2 and 3 Centrifuge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-269-325 Supplier HemoCue America Supplier No. SV05
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with the StatSpin VET Express Primary Tube Centrifuge
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2mL BD Microtainer Tubes; Express 2 and 3 Centrifuge
Product Type Adapter