Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America StatSpin™ VT Multipurpose Veterinary Centrifuge

Catalog No. 22269521
Change view
Click to view available options
Rotor Included:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Rotor Included
22-269-521 Yes
22-269-520 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-269-521 Supplier HemoCue America Supplier No. SSVT1

missing translation for 'validMainframeMsg'

  • Compact, less than 7 in. (17.8cm) of bench space
  • Quiet operation
  • Separates serum or plasma in 30 to 90 seconds, hematocrit in two minutes, and urine sediment in 45 seconds
  • Preprogrammed run cycles
  • Membrane touch panel and microprocessor control
  • Easy-to-clean, coated steel interior
  • Remote power supply for installation flexibility
  • Cover interlock system for added safety
  • Accepts accessories from previous StatSpin models (StatSpin 1, 2, 3, 4; StatSpin MP, RP, CritSpin™)

Compliance

UL listed, CE marked

Warranty and Services

One-year warranty from date of installation; extended warranties available upon request

TRUSTED_SUSTAINABILITY

Specifications

Max. RCF 13,700 x g
Max. Speed 15,800 rpm
Refrigerated No
Profile (Acceleration/Braking) Approx. 6 sec./<15 sec.
Electrical Requirements 100/240 V 50/60 Hz
Certifications/Compliance UL listed; CE
Height (English) Exterior 6 in.
Height (Metric) Exterior 15.2 cm
Length (English) Exterior 8.6 in.
Length (Metric) Exterior 21.9 cm
Width (English) Exterior 6.6 in.
Width (Metric) Exterior 16.8 cm
Controller Type Microprocessor
Rotor Included Yes
Dimensions (HWD) 6 x 6.6 x 8.6 in. (15.2 x 16.8 x 21.9 cm)
Net Weight 3.75 lb. (1.7 kg)
Program Storage Programmed Cycles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.