Learn More
Medchemexpress LLC HY-13818 10mg Medchemexpress, Stattic CAS:19983-44-9 Purity:>98%
SDP
Catalog No. 502029034
Encompass_Preferred
Medchemexpress, HY-13818 10mg Stattic CAS:19983-44-9 Stattic is a potent STAT3 inhibitor. Stattic inhibits STAT3 phosphorylation (at Y705 and S727). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9034 Supplier Medchemexpress LLC Supplier No. HY1381810MG
Medchemexpress, HY-13818 10mg Stattic CAS:19983-44-9 Stattic is a potent STAT3 inhibitor. Stattic inhibits STAT3 phosphorylation (at Y705 and S727). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.