Learn More
Apexbio Technology LLC Stattic 19983-44-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A22245.1
Encompass_Preferred
Stattic (CAS 19983-44-9) is a small-molecule inhibitor specifically targeting signal transducer and activator of transcription 3 (STAT3) It suppresses STAT3 activation by blocking its dimerization nuclear translocation and subsequent transcriptional activity In vitro studies demonstrate Stattic inhibits proliferation in multiple head and neck squamous cell carcinoma (HNSCC) cell lines including UM-SCC-17B OSC-19 Cal33 and UM-SCC-22B with reported IC50 values ranging from approximately 2 3 to 3 5 M Additionally Stattic downregulates STAT3-mediated hypoxia-inducible factor-1 (HIF-1) expression resulting in enhanced radiosensitivity in HNSCC cells and shows activity in mouse xenograft tumor models Therefore Stattic serves as a research tool for investigating STAT3-associated signaling pathways and cancer biology
Catalog No. 50-000-61425