Learn More
Apexbio Technology LLC Stattic, 19983-44-9, MFCD00173799, 25 mg
SDP
Supplier:  Apexbio Technology LLC A222425
Encompass_Preferred
MF: C8H5NO4S, MW: 211.19, Solubility: 10.6mg/mL in DMSO. Stattic is a small molecule inhibitor of STAT3. Stattic is reported to selectively inhibit dimerization, activation and nuclear translocation of STAT3. It can also induce the apoptosis of STAT3-dependent breast cancer cell lines.
Catalog No. 50-101-0965