Learn More
Abcam Stattic, STAT3 inhibitor, 50MG
Catalog No. 502707095
Encompass_Preferred
MW 211.2 Da, Purity >99%. Selective STAT3 inhibitor. Inhibits activation, dimerization and nuclear translocation of STAT3 by interacting with the SH2 domain. Displays selectivity over STAT1, STAT5, c-Myc/Max, Jun/Jun and Lck. Induces apoptosis in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7095 Supplier Abcam Supplier No. AB12095250MG
MW 211.2 Da, Purity >99%. Selective STAT3 inhibitor. Inhibits activation, dimerization and nuclear translocation of STAT3 by interacting with the SH2 domain. Displays selectivity over STAT1, STAT5, c-Myc/Max, Jun/Jun and Lck. Induces apoptosis in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement