Learn More
LifeSign Status BTA Urine Test Kit
SDP
Catalog No. 23264798 Shop All Lifesign LLC Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-264-798 25 Tests
1 options
Catalog No. 23-264-798 Supplier LifeSign Supplier No. 39025
Status BTA is a rapid test for the qualitative detection of bladder tumor associated antigen in human urine.
The test is indicated for use as an aid in the management of bladder cancer patients in conjunction with cystoscopy. The simple urine sample collection also allows for improved patient comfort. Because tumor markers can be used to assess the response of a tumor treatment and for prognosis, rapid screening tests like the Status BTA product are very helpful in the treatment management of cancer patients.
  • CLIA Waived
  • Results available in 5 minutes
  • Simple procedure requires only adding 3 drops of urine to test device; no sample preparation needed
  • Accurate test results with excellent sensitivity and specificity
  • Uses monoclonal antibodies to detect the presence of bladder tumor associated antigen in urine
  • Detects early state recurrance cancer that cytology may miss
  • Adjunct test to cystoscopy
The test is indicated for use as an aid in the management of bladder cancer patients in conjunction with cystoscopy.  The simple urine sample collection also allows for improved patient comfort.  Because tumor markers can be used to assess the response of a tumor treatment and for prognosis, rapid screening tests like the Status BTA product are very helpful in the treatment management of cancer patients.
  • CLIA Waived
  • Results available in 5 minutes
  • Simple procedure requires only adding 3 drops of urine to test device; no sample preparation needed
  • Accurate test results with excellent sensitivity and specificity
  • Uses monoclonal antibodies to detect the presence of bladder tumor associated antigen in urine
  • Detects early state recurrance cancer that cytology may miss
  • Adjunct test to cystoscopy
Specifications
Detectable Analytes Bladder tumor associated antigen
Quantity 25 Tests
Test Time 5 min.