Learn More
LifeSign Status hCG Strip Test Kit
SDP
Catalog No. 23264794 Shop All Lifesign LLC Products
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-264-794 25 Tests
1 options
Catalog No. 23-264-794 Supplier LifeSign Supplier No. 35235
Rapid immunoassay strip test for the detection of human chorionic gonadotropin in urine
Features:
  • Dip and read simplicity for ease of use
  • 25mIU/mL sensitivity for early pregnancy detection
  • Compact strip size, reduces biohazard disposal
  • Results in 3 to 5 minutes
  • CLIA waived
Specifications
Detectable Analytes Human chorionic gonadotropin
Format Strip
Quantity 25 Tests