Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Status hCG Test Kit
SDP

Catalog No. 23264793 Shop All Lifesign LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Sample Type:
Urine
Urine, Serum
Clia Complexity:
Waived
Waived (Urine)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Clia Complexity
23-264-793 Urine Waived
23-264-792 Urine, Serum Waived (Urine)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-264-793 Supplier LifeSign Supplier No. 35135
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid immunoassay test for the detection of human chorionic gonadotropin in urine

Features:

  • Excellent sensitivity, 25mIU/mL, regardless of specimen for early pregnancy detection
  • Clear, definitive result lines are easy to read
  • Simple one-step procedure
  • Results in 3 to 5 minutes
  • CLIA waived for urine

Specifications

Clia Complexity Waived
Detectable Analytes Human chorionic gonadotropin
Format Cassette
Sample Type Urine
Sensitivity 25 mIU/mL
Quantity 35 Tests
Test Time 3 to 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.