Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LifeSign Status Strep A Test Kit
SDP

Catalog No. 23264790 Shop All Lifesign LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Tests
30 Tests
Format:
Flip-design Cassette
Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
23-264-790 30 Tests Strip
23-264-789 25 Tests Flip-design Cassette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-264-790 Supplier LifeSign Supplier No. 34130
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid immunochromatographic assay for the detection of group A streptococcal antigen directly from throat swab specimens

Specifications

Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Format Strip
Sample Type Throat Swab
Sensitivity 96.2%
Type Immunochromatographic Assay
Includes External negative and positve contols
Quantity 30 Tests
Specificity 0.987
Test Time 5 to 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.