Learn More
MALIX LLC For The Qualitative Detection of Infectious Mononucleosis Heterophile Antibodies in Whole Blood, Serum, or Plasma
Catalog No. NC3235775
Encompass_Preferred
StatusFirst Mono Control Set - POS & NEG (2 x 0.5 ml)
CLIA WAIVED for whole blood
FDA cleared with no age restrictions
Single reagent procedure provides results in 3-8 minutes
Fingerstick whole blood convenience
Superior 99.9% sensitivity for earlier IM detection, specificity of 98.8%New plastic sample transfer pipettes
Catalog No. NC3235775 Supplier MALIX LLC Supplier No. STATMONPN
StatusFirst Mono Control Set - POS & NEG (2 x 0.5 ml)
CLIA WAIVED for whole blood
FDA cleared with no age restrictions
Single reagent procedure provides results in 3-8 minutes
Fingerstick whole blood convenience
Superior 99.9% sensitivity for earlier IM detection, specificity of 98.8%New plastic sample transfer pipettes