Learn More
Aviva Systems Biology Corp STAU1 Antibody - N-terminal region (ARP40537_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40537_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against STAU1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human STAU1. Alternate names: STAUApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57513