Learn More
Aviva Systems Biology Corp STAU1 Antibody - N-terminal region (ARP40860_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40860_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAU1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STAU1. Alternate names: FLJ25010, STAUApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-57642