Learn More
STEMCELL Technologies Stauprimide, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 72652
Encompass
Stauprimide specifically inhibits the nuclear localization of NME2, which results in the suppression of c-MYC—a key regulator of pluripotency—thereby priming cells for differentiation (Zhu et al.).DIFFERENTIATION· Enhances cytokine-mediated directed differentiation of mouse and human pluripotent stem cells to multiple lineages, including definitive endoderm, neural progenitors, and mesodermal derivatives such as cardiomyocytes (Zhu et al.; Tahamtani et al.).
Catalog No. 50-197-5956