Learn More
Cayman Chemical StaurosporIn 10mg
SDP
Catalog No. 502894618
Encompass_Preferred
A potent inhibitor of PKC (IC50 = 2.7 nM for the rat brain enzyme)
Catalog No. 50-289-4618 Supplier Cayman Chemical Supplier No. 8159010 MG
A potent inhibitor of PKC (IC50 = 2.7 nM for the rat brain enzyme)