Learn More
Alkali Scientific Staurosporine, 1 Mg
SDP
Catalog No. 5000661086
Encompass_Preferred
Staurosporine, 1 mg, is a powerful protein kinase inhibitor that is commonly used in scientific research to study cell signaling, apoptosis, and disease mechanisms. Available in powder form, it is often reconstituted for use in cell culture experiments or animal models. Staurosporine acts by inhibiting a variety of protein kinases that regulate cellular processes such as growth, differentiation, and survival. This makes it an essential tool in the study of diseases like cancer, cardiovascular conditions, and neurological disorders. The 1mg vial format is used in more extensive experimental protocols where higher concentrations are needed for effective inhibition of kinase activity.
Catalog No. 50-006-61086 Supplier Alkali Scientific Supplier No. STA003CP
Staurosporine, 1 mg, is a powerful protein kinase inhibitor that is commonly used in scientific research to study cell signaling, apoptosis, and disease mechanisms. Available in powder form, it is often reconstituted for use in cell culture experiments or animal models. Staurosporine acts by inhibiting a variety of protein kinases that regulate cellular processes such as growth, differentiation, and survival. This makes it an essential tool in the study of diseases like cancer, cardiovascular conditions, and neurological disorders. The 1mg vial format is used in more extensive experimental protocols where higher concentrations are needed for effective inhibition of kinase activity.