Learn More
Research Products International Corp Staurosporine, 100 Micrograms
SDP
Catalog No. 501368601
Encompass_Preferred
Staurosporine is a potent natural compound and is a crucial tool in studying signal transduction pathways, cell cycle regulation, and cancer therapy. In cell biology, Staurosporine is widely used to investigate cellular processes involving protein kinases. It acts as a broad-spectrum kinase inhibitor, targeting various protein kinases involved in signal transduction, which helps researchers explore complex intracellular signaling networks. This has far-reaching implications for understanding the mechanisms underlying cell growth, differentiation, and apoptosis.
Pharmacologists and pharmaceutical researchers leverage Staurosporine in drug discovery and development. Its ability to inhibit kinases makes it valuable in identifying potential therapeutic targets and assessing the efficacy of novel compounds for treating diseases like cancer. In cancer research.
Catalog No. 50-136-8601 Supplier Research Products International Corp Supplier No. S635000.0001
Staurosporine is a potent natural compound and is a crucial tool in studying signal transduction pathways, cell cycle regulation, and cancer therapy. In cell biology, Staurosporine is widely used to investigate cellular processes involving protein kinases. It acts as a broad-spectrum kinase inhibitor, targeting various protein kinases involved in signal transduction, which helps researchers explore complex intracellular signaling networks. This has far-reaching implications for understanding the mechanisms underlying cell growth, differentiation, and apoptosis.
Pharmacologists and pharmaceutical researchers leverage Staurosporine in drug discovery and development. Its ability to inhibit kinases makes it valuable in identifying potential therapeutic targets and assessing the efficacy of novel compounds for treating diseases like cancer. In cancer research.