Learn More
Biotium Staurosporine, 100ug
Catalog No. 501964055
Encompass_Preferred
Staurosporine is a potent and cell-permeable inhibitor of a wide variety of protein kinases including protein kinase C (IC50 = 0.7 nM), protein kinase A (IC50 = 7 nM), and protein kinase G (IC50 = 8.5 nM). Staurosporine induces apoptosis in human neuroblastoma cells and chick embryonic neurons. Properties: Soluble in DMSO or ethanol; Store at -20°C; C28H26N4O3; MW: 466.20; [62996-74-1]
Catalog No. 50-196-4055 Supplier Biotium Supplier No. 00025
Staurosporine is a potent and cell-permeable inhibitor of a wide variety of protein kinases including protein kinase C (IC50 = 0.7 nM), protein kinase A (IC50 = 7 nM), and protein kinase G (IC50 = 8.5 nM). Staurosporine induces apoptosis in human neuroblastoma cells and chick embryonic neurons. Properties: Soluble in DMSO or ethanol; Store at -20°C; C28H26N4O3; MW: 466.20; [62996-74-1]