Learn More
Abcam Staurosporine, Protein kinase inhibitor, 100UG
Supplier:  Abcam AB120056100UG
Encompass_Preferred
MW 466.5 Da, Purity >98%. Potent, cell-permeable, broad spectrum inhibitor of protein kinases. Kinases inhibited include: protein kinase C (IC₅₀ = 3 nM), cAMP-dependent protein kinase (IC₅₀ = 8 nM) and p60v-src (IC₅₀ = 6 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0748115