Learn More
Medchemexpress LLC HY-15141 2mg Medchemexpress, Staurosporine CAS:62996-74-1 Purity:>98%
SDP
Catalog No. 502029080
Encompass_Preferred
Medchemexpress, HY-15141 2mg Staurosporine CAS:62996-74-1 Staurosporine is a potent and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine is an apoptosis inducer[2][3][4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9080 Supplier Medchemexpress LLC Supplier No. HY151412MG
Medchemexpress, HY-15141 2mg Staurosporine CAS:62996-74-1 Staurosporine is a potent and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine is an apoptosis inducer[2][3][4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.