Learn More
Selleck Chemical LLC Staurosporine 2mg 62996-74-1 CGP 41251
SDP
Catalog No. NC0535441
Encompass_Preferred
Staurosporine (CGP 41251) is a potent PKC inhibitor for PKC?, PKC? and PKC? with IC50 of 2 nM, 5 nM and 4 nM, less potent to PKC? (20 nM), PKC? (73 nM) and little active to PKC? (1086 nM) in cell-free assays. Also shows inhibitory activities on other kinases, such as PKA, PKG, S6K, CaMKII, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0535441 Supplier Selleck Chemical LLC Supplier No. S14212MG
Staurosporine (CGP 41251) is a potent PKC inhibitor for PKC?, PKC? and PKC? with IC50 of 2 nM, 5 nM and 4 nM, less potent to PKC? (20 nM), PKC? (73 nM) and little active to PKC? (1086 nM) in cell-free assays. Also shows inhibitory activities on other kinases, such as PKA, PKG, S6K, CaMKII, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.