Learn More
Alkali Scientific Staurosporine, 500 Micrograms
SDP
Catalog No. 5000661152
Encompass_Preferred
Staurosporine, 500 micrograms, is a potent protein kinase inhibitor used in cell biology and pharmacological research. Available in powder form, it is commonly used to study cellular signaling pathways and apoptosis mechanisms. Staurosporine works by inhibiting a broad range of protein kinases, including those involved in cell division, differentiation, and apoptosis. This makes it a useful tool in studying cancer, neurodegenerative diseases, and other conditions related to abnormal cell growth and survival. The 500-microgram vial is suitable for small-scale laboratory experiments, providing precise dosing for kinase inhibition studies and cell death research.
Catalog No. 50-006-61152 Supplier Alkali Scientific Supplier No. STA002CP
Staurosporine, 500 micrograms, is a potent protein kinase inhibitor used in cell biology and pharmacological research. Available in powder form, it is commonly used to study cellular signaling pathways and apoptosis mechanisms. Staurosporine works by inhibiting a broad range of protein kinases, including those involved in cell division, differentiation, and apoptosis. This makes it a useful tool in studying cancer, neurodegenerative diseases, and other conditions related to abnormal cell growth and survival. The 500-microgram vial is suitable for small-scale laboratory experiments, providing precise dosing for kinase inhibition studies and cell death research.