Learn More
AdipoGen Staurosporine, 62996-74-1, MFCD18252446, 5 mg
SDP
Catalog No. 501146222
Encompass_Preferred
MF: C28H26N4O3, MW: 466.5, ≥98%, Appearance: Off-white to yellow solid. Antibiotic. Antifungal and anti-yeast activity. Inhibits platelet aggregation induced by collagen or ADP. Has no effect on thrombin-induced platelet aggregation. Potent, cell permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases. Inhibits protein kinase A, CaM kinase, myosin light chain kinase, protein kinase C, protein kinase G, CDK1/cyclin B, CDK2/cyclin A, CDK4/cyclin D, CDK5/p25, GSK-3beta and Pim-1 kinase. Key apoptosis inducer. Topoisomerase II (Topo II) inhibitor.
Catalog No. 50-114-6222 Supplier AdipoGen Supplier No. AGCN20022M005
MF: C28H26N4O3, MW: 466.5, ≥98%, Appearance: Off-white to yellow solid. Antibiotic. Antifungal and anti-yeast activity. Inhibits platelet aggregation induced by collagen or ADP. Has no effect on thrombin-induced platelet aggregation. Potent, cell permeable, reversible, ATP-competitive and broad spectrum inhibitor of protein kinases. Inhibits protein kinase A, CaM kinase, myosin light chain kinase, protein kinase C, protein kinase G, CDK1/cyclin B, CDK2/cyclin A, CDK4/cyclin D, CDK5/p25, GSK-3beta and Pim-1 kinase. Key apoptosis inducer. Topoisomerase II (Topo II) inhibitor.