Learn More
Lkt Laboratories CAS 62996-74-1, ≥98%, PKC and mammalian RNA splicing inhibitor produced by Streptomyces. It is a precursor in the synthesis of K252c.
SDP
Catalog No. NC2539840
Encompass_Preferred
Staurosporine is an alkaloid initially produced by Streptomyces that inhibits PKC and other protein kinases. Staurosporine is a precursor in the synthesis of kinase inhibitors K252c and PKC412. This compound exhibits anticancer, anti-parasitic, and anti-protozoan activities. Staurosporine induces apoptosis in hepatocarcinoma cells by decreasing levels of p38 MAPK, pERK, and DNA methyltransferase 3B (DNMT 3B) and increasing levels of pJNK. Staurosporine also causes cell death in Trypanosoma. This compound also inhibits mammalian RNA splicing.
Catalog No. NC2539840 Supplier Lkt Laboratories Supplier No. S7600
Staurosporine is an alkaloid initially produced by Streptomyces that inhibits PKC and other protein kinases. Staurosporine is a precursor in the synthesis of kinase inhibitors K252c and PKC412. This compound exhibits anticancer, anti-parasitic, and anti-protozoan activities. Staurosporine induces apoptosis in hepatocarcinoma cells by decreasing levels of p38 MAPK, pERK, and DNA methyltransferase 3B (DNMT 3B) and increasing levels of pJNK. Staurosporine also causes cell death in Trypanosoma. This compound also inhibits mammalian RNA splicing.