Learn More
Selleck Chemical LLC Staurosporine (STS) S1421-25mg
SDP
Catalog No. 503108339
Encompass_Preferred
Staurosporine (STS) is a potent PKC inhibitor for PKC PKC and PKC with IC50 of 2 nM 5 nM and 4 nM less potent to PKC (20 nM) PKC (73 nM) and little active to PKC (1086 nM) in cell-free assays Also shows inhibitory activities on other kinases such as PKA PKG S6K CaMKII etc Phase 3
Catalog No. 50-310-8339 Supplier Selleck Chemical LLC Supplier No. S142125MG
Staurosporine (STS) is a potent PKC inhibitor for PKC PKC and PKC with IC50 of 2 nM 5 nM and 4 nM less potent to PKC (20 nM) PKC (73 nM) and little active to PKC (1086 nM) in cell-free assays Also shows inhibitory activities on other kinases such as PKA PKG S6K CaMKII etc Phase 3