Learn More
Sigma Aldrich Fine Chemicals Biosciences Staurosporine Staurosporin
Catalog No. 502905587
Encompass
Suitable for use as a potent inhibitor of PKC and other protein kinases in vitro and in cultured cells.
Catalog No. 50-290-5587 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 19123MG
Add to Cart
Add to Cart
Suitable for use as a potent inhibitor of PKC and other protein kinases in vitro and in cultured cells.