Learn More
Cayman Chemical StavudIn-13C-d3 500ug
SDP
Catalog No. 502937488
Encompass_Preferred
An internal standard for the quantification of stavudine by GC- or LC-MS
Catalog No. 50-293-7488 Supplier Cayman Chemical Supplier No. 28887500 UG
An internal standard for the quantification of stavudine by GC- or LC-MS