Learn More
Abcam StayRed/AP 30ml (Alcohol & x ylene Compatible), 30ML
Catalog No. 502585560
Encompass_Preferred
StayRed/AP is a substrate chromogen system designed to be used for either IHC or ISH when using alkaline phosphate detection.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5560 Supplier Abcam Supplier No. AB10374130ML
StayRed/AP is a substrate chromogen system designed to be used for either IHC or ISH when using alkaline phosphate detection.
The product is subject to the following: Abcam Restricted Use Statement