Learn More
UNITRON Stays lens, 10-Diopter
SDP
Catalog No. 01189337 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Color:
White
Catalog No. Color
01-189-337 White
1 options
Catalog No. 01-189-337 Supplier UNITRON Supplier No. SPD026119
Add to Cart
Add to Cart
Engineered with a precision cup design for a secure fit, enabling easy application and enhanced visibility for detailed inspection tasks.
  • Provides 3.5X magnification (10 diopter) for enhanced detail and accuracy during close-up work.
  • Precision cup design ensures a stable and secure fit for consistent viewing performance.
  • Working distance of 2.9 in. with 3D/3.5 D lens and 2.6 in. with 5D lens for versatile application.
  • Compact 2 in. lens diameter (2.5 in. including silicone gasket frame) for controlled and precise viewing.
Specifications
Type Lens
Autoclavable Not Autoclavable
Color White
Sterility Non-sterile