Learn More
Abcam StayYellow/HRP, 30ML
Catalog No. 502582580
Encompass_Preferred
StayYellow/HRP is a substrate chromogen system designed to be used for either IHC or ISH when using HRP detection.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2580 Supplier Abcam Supplier No. AB16956130ML
StayYellow/HRP is a substrate chromogen system designed to be used for either IHC or ISH when using HRP detection.
The product is subject to the following: Abcam Restricted Use Statement