Learn More
Sigma Aldrich Fine Chemicals Biosciences Penicillin-Streptomycin, 100 mL
Catalog No. NC0363640
Encompass
Supplement in Dulbecco′s modified Eagle′s medium (DMEM) to maintain the Huh7 liver hepatoma cell line. Supplement in Roswell park memorial institute-1640 (RPMI) medium to maintain mouse lymphoma YAC-1 cells. A component in minimum essential medium Eagle to treat individual mouse spleens for the preparation of splenocytes.
Catalog No. NC0363640 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P4458100ML
Supplement in Dulbecco′s modified Eagle′s medium (DMEM) to maintain the Huh7 liver hepatoma cell line. Supplement in Roswell park memorial institute-1640 (RPMI) medium to maintain mouse lymphoma YAC-1 cells. A component in minimum essential medium Eagle to treat individual mouse spleens for the preparation of splenocytes.