Learn More
Foxx Life Sciences EZBio Connector Assembly 3/4" x 3/4" Steam-Thru (STC2020900) with AseptiquikHT S (AQS33012HT) Series Connector, Sterile, 5/CS
Catalog No. 502551723
Encompass_Preferred
The AseptiQuik STC Connectors allow a quick sterile connection between biopharmaceutical processing equipment and single-use systems. The Union of the two connectors into a single unit through a Foxx sanitary Tri-clamp allows AseptiQuik sterile connection to be steamed onto the stainless steel equipment via SIP. After the SIP cycle, a wide range of Single-Use Systems can be connected with the AseptiQuik G or AseptiQuik S Sterile connector. The SIP process can be done in advance, allowing a quick and easy sterile connection to the AseptiQuik half without waiting 30-60 minutes for SIP before harvest.
The AseptiQuik STC Connector connects stainless steel processing equipment via 3/4"" or 1-1/2"" Sanitary Termination.
Steam Position
Temperature: 39°F to 104°F (4°C to 40°C)
Pressure: Up to 35 psi, 2.4 bar (Steam-Thru II)
Flow Position
Temperature: 39°F to 104°F (4°C to 40°C)
Pressure: Vacuum to 20 psi, 1.4 bar
Catalog No. 50-255-1723 Supplier Foxx Life Sciences Supplier No. 446350DFLS
The AseptiQuik STC Connectors allow a quick sterile connection between biopharmaceutical processing equipment and single-use systems. The Union of the two connectors into a single unit through a Foxx sanitary Tri-clamp allows AseptiQuik sterile connection to be steamed onto the stainless steel equipment via SIP. After the SIP cycle, a wide range of Single-Use Systems can be connected with the AseptiQuik G or AseptiQuik S Sterile connector. The SIP process can be done in advance, allowing a quick and easy sterile connection to the AseptiQuik half without waiting 30-60 minutes for SIP before harvest.
The AseptiQuik STC Connector connects stainless steel processing equipment via 3/4"" or 1-1/2"" Sanitary Termination.
Steam Position
Temperature: 39°F to 104°F (4°C to 40°C)
Pressure: Up to 35 psi, 2.4 bar (Steam-Thru II)
Flow Position
Temperature: 39°F to 104°F (4°C to 40°C)
Pressure: Vacuum to 20 psi, 1.4 bar