Learn More
Aviva Systems Biology Corp STC2 Antibody - C-terminal region (ARP64151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STC2. It was validated on Western Blot. . Alternate names: STC-2, STCRPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-191