Learn More
Research Products International Corp Stacking Buffer 4X Solution 1L
SDP
Catalog No. 502230485
Encompass_Preferred
4X concentrated buffer solution for preparation of Laemmli SDS-PAGE resolving gels.
Catalog No. 50-223-0485 Supplier Research Products International Corp Supplier No. S282001000.0
4X concentrated buffer solution for preparation of Laemmli SDS-PAGE resolving gels.