Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ 2000 ppm: 2-Bromo-1-chloropropane, Fluorobenzene in Methanol ASTM D5580, Ricca Chemical
SDP

Catalog No. RV0190341R Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV0190341R 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV0190341R Supplier Ricca Chemical Company Supplier No. RV0190341R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Specifications

Chemical Name or Material VeriSpec™ Std 2000 ppm: 2-Bromo-1-Chloropropane, Fluorobenzene in Methanol
Name Note ASTM D5580
Packaging Ampule
CAS 67-56-1, 462-06-6, 3017-95-6
CAS Min % 99.6, 0.2, 0.2
CAS Max % 99.6, 0.2, 0.2
Density 0.8
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Multi-element Standard Solution
Quantity 1 mL
Grade High Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.