Learn More
Perkin Elmer US LLC Alternate Interferents B, 5% HNO3/trace Tartaric Acid/trace HF, 125 mL
Catalog No. 509045745
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 5% HNO3 100 µg/mL: Cd, Ni, Zn 60 µg/mL: Sb 50 µg/mL: Ba, Be, Co, Cr, Cu, Mn, V 20 µg/mL: Ag 10 µg/mL: As, Tl 5 µg/mL: Pb, Se
Catalog No. 50-904-5745 Supplier Perkin Elmer US LLC Supplier No. N9300227
Add to Cart
Add to Cart
Concentration of Element includes: Matrix Concentration 5% HNO3 100 µg/mL: Cd, Ni, Zn 60 µg/mL: Sb 50 µg/mL: Ba, Be, Co, Cr, Cu, Mn, V 20 µg/mL: Ag 10 µg/mL: As, Tl 5 µg/mL: Pb, Se